Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human A2LD1 His Protein
SDP

Catalog No. NBP17879950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17879950 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17879950 Supplier Novus Biologicals™ Supplier No. NBP17879950UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-153 of Human A2LD1 The Recombinant Human A2LD1 Protein is derived from E. coli. The Recombinant Human A2LD1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q9BVM4
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0) containing 10% glycerol, 0.1M NaCl
Gene ID (Entrez) 87769
Molecular Weight (g/mol) 19.4kDa
Name A2LD1 Protein
Quantity 50 μg
Source E. Coli
Immunogen A2LD1, 1-153aa, Human, His tag, E.coli (NP_149101).
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.