Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human ACE-2 Fc Chimera Protein
Click to view available options
Quantity:
100 μg
20 μg
Description
Measured by its binding ability in a functional ELISA with Recombinant Viral SARS-CoV-2 S Protein RBD Fc Chimera (Catalog # 10499-CV ). Measured by its ability to cleave a fluorogenic peptide substrate, Mca-YVADAPK(Dnp)-OH (Catalog # ES007 ). The specific activity is >550 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | Q9BYF1.2 |
| For Use With (Application) | Bioactivity, Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, ZnCl2 and Glycerol. |
| Gene ID (Entrez) | 59272 |
| Molecular Weight (g/mol) | M.W.-observed: 124-130 kDa, under reducing conditions, M.W.-theoretical: 110 kDa |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human ACE-2 protein Human ACE-2 (Gln18-Ser740) (N-terminus) Accession # Q9BYF1.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction