Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human ACSF2 His Protein
SDP

Catalog No. NBP25216601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25216601 0.1 mg
NBP25216605 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25216601 Supplier Novus Biologicals™ Supplier No. NBP2521660.1MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 42-615 of Human ACSF2 The Recombinant Human ACSF2 Protein is derived from E. coli. The Recombinant Human ACSF2 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline pH 7.4 containing 10% glycerol
Gene ID (Entrez) 80221
Molecular Weight (g/mol) TMW: 66.1kDa
Quantity 0.1 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias ACSMW, acyl-CoA synthetase family member 2, AVYV493, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.26, mitochondrial, PPARG binding, long chain fatty acid acyl Co-A ligase like
Common Name ACSF2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.