Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Active FAK Protein
SDP

Catalog No. 4467KS010 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4467KS010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4467KS010 Supplier R&D Systems™ Supplier No. 4467KS010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The specific activity of FAK was determined to be 51 nmol/min/mg as per Activity Assay Protocol, and was equivalent to 291.5 nmol/min/mg as per radiometric assay.

Specifications

Components Supplied in 50mM sodium phosphate (pH 7.0), 300mM NaCl, 0.25mM DTT, 150mM imidazole, 0.1mM PMSF, and 25% glycerol.
Purity >75%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.)
For Use With (Application) Bioactivity
Storage Buffer Supplied in 50 mM sodium phosphate (pH 7.0), 300 mM NaCl, 150 mM Imidazole, 0.1 mM PMSF, 0.25 mM DTT, and 25% Glycerol.
Content And Storage This product is stable at <= -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store (at recommended temperature. Avoid repeated freeze-thaw cycles.)
Source Spodoptera frugiperda, Sf 9 (baculovirus)-derived human FAK protein
Quantity 10 μg
Product Type Recombinant Human Focal adhesion kinase 1
Molecular Weight (g/mol) Observed MW: 35kDa
Product Line R&D Systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.