Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Active p38 alpha Protein
SDP

Catalog No. 5477KS010 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5477KS010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 5477KS010 Supplier R&D Systems™ Supplier No. 5477KS010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The activity of p38 alpha is typically 148-201 nmol/min/mg using an ATF2 substrate (see activity assay protocol).

Specifications

Components Supplied in 50mM Tris-HCl (pH 7.5), 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, and 25% glycerol.
Purity >70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
For Use With (Application) Bioactivity
Storage Buffer Supplied in 50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Content And Storage This product is stable at <= -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store (at recommended temperature. Avoid repeated freeze-thaw cycles.)
Accession Number NM_139012
Gene ID (Entrez) 1432
Gene Symbol MAPK14
Source Spodoptera frugiperda,Sf 9 (baculovirus)-derived human p38 alpha protein Accession &Num; NM_139012
Quantity 10 μg
Product Type Recombinant Human Active p38 alpha, CF
Molecular Weight (g/mol) M.W. (Observed): 67 kDa
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.