Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Activin AB Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with Trehalose.
Description
The Recombinant Human Activin AB Protein is derived from CHO. The Recombinant Human Activin AB Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
Molecular Weight (g/mol) | 12.8 kDa (betaA subunit) and 13 kDa (betaB subunit) |
Name | Activin AB |
Quantity | 5 μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human Activin AB protein, (N-terminus) Human Activin beta A (Gly311-Ser426) Accession &Num; NP_002183.1 Human Activin beta B (Gly293-Ala407) Accession &Num; NP_002184.2 (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked heterodimer |
Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction