Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human ADAMTS13 (Full Length) Protein
Click to view available options
Quantity:
20 μg
Description
The Recombinant Human ADAMTS13 (Full Length) Protein is derived from CHO. The Recombinant Human ADAMTS13 (Full Length) Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Formulation | Supplied as a 0.2μm filtered solution in HEPES and NaCl. |
Gene ID (Entrez) | 11093 |
Molecular Weight (g/mol) | Theoretical Molecular weight:147 kDa & 151 kDa |
Name | ADAMTS13 Full Length |
Quantity | 20 μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human ADAMTS13 protein Gln34-Thr1427,with a C-terminal 10-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction