Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human ALDH1A2 His-tag
Click to view available options
Quantity:
20 μg
Description
- Recombinant Protein
- Carrier Free (CF)
- The carrier free version does not contain Bovine Serum Albumin (BSA)
Specifications
Specifications
| Accession Number | O94788-1 |
| For Use With (Application) | Enzyme Activity |
| Gene ID (Entrez) | 8854 |
| Molecular Weight (g/mol) | 58 kDa |
| Purification Method | >95%, by SDS-PAGE under reducing conditions and visualized by silver stain |
| Quantity | 20 μg |
| Source | E. coli-derived human ALDH1A2 protein Thr2-Ser518 with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70°C as supplied. 3 months, -20 to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction