Learn More
Novus Biologicals™ Recombinant Human alpha-Synuclein Active, Pre-formed Fibrils, (Type 2) Protein
Shop All R&D Systems Products

Description
- Thioflavin T emission curve shows only a small increase in fluorescence (indicative of alpha synuclein aggregation) when Type 2 alpha synuclein PFFs (NBP2-54787) are combined with alpha synuclein monomers (NBP2-54788 or NBP2-54786).
- Certain biological activities in other neuronal cells cannot be ruled out.
- Researchers should test compatibility prior to use.
Specifications
Specifications
| For Use With (Application) | In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot |
| Gene ID (Entrez) | 6622 |
| Quantity | 0.1 mg |
| Source | E. coli |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles |
| Species | Human |
| Buffer | PBS pH 7.4 |
| Purity or Quality Grade | Ion exchange chromatography |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.