Learn More
Novus Biologicals Recombinant Human alpha-Synuclein Oligomers, (Dopamine HCL Stabilized) Protein
Shop All R&D Systems Products

Specifications
Specifications
| For Use With (Application) | In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot |
| Formulation | PBS pH 7.4 |
| Gene ID (Entrez) | 6622 |
| Quantity | 2 x 100 μg Vials |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Gene Alias | alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein |
| Gene Symbol | SNCA |
| Research Category | Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience |
| Purity or Quality Grade | >95%, by SDS-PAGE |
| Protein | alpha-Synuclein |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.