Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Recombinant Human AlphaB Crystallin/CRYAB His Protein
SDP

Catalog No. NB355228 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355228 250 μg
NB355227 50 μg
NB355229 20 μg
NB355230 10 μg
NB355231 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. NB355228 Supplier Novus Biologicals Supplier No. NBP324599250UG
Add to Cart
Add to Cart

Specifications

Concentration 1 mg/ml
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 1410
Molecular Weight (g/mol) 21.2 kDa
Quantity 250 μg
Storage Requirements Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Gene Alias alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component
Gene Symbol CRYAB
Research Category Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision
Purity or Quality Grade >90%, by SDS-PAGE
Protein AlphaB Crystallin/CRYAB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.