Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human AlphaB Crystallin/CRYAB Protein
SDP

Catalog No. NBC118352 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118352 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118352 Supplier Novus Biologicals™ Supplier No. NBC118352
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-175 of Human AlphaB Crystallin/CRYAB The Recombinant Human AlphaB Crystallin/CRYAB Protein is derived from E. coli. The Recombinant Human AlphaB Crystallin/CRYAB Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001876.1
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5), 50mM NaCl, 1mM EDTA
Gene ID (Entrez) 1410
Molecular Weight (g/mol) 20.1kDa
Name AlphaB Crystallin/CRYAB protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MDIAIHHPWIRRPFFPFHSPSRLFDQFFGEHLLESDLFPTSTSLSPFYLRPPSFLRAPSWFDTGLSEMRLEKDRFSVNLDVKHFSPEELKVKVLGDVIEVHGKHEERQDEHGFISREFHRKYRIPADVDPLTITSSLSSDGVLTVNGPRKQVSGPERTIPITREEKPAVTAAPKK
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.