Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Angiopoietin-2, Biotinylated Protein
Click to view available options
:
10μg; Biotin
Description
The Recombinant Human Angiopoietin-2, Biotinylated Protein is derived from NS0. The Recombinant Human Angiopoietin-2, Biotinylated Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Accession Number | O15123 |
For Use With (Application) | Bioactivity |
Gene ID (Entrez) | 285 |
Molecular Weight (g/mol) | Theoretical M.W.: 56kDa (unlabeled) |
Quantity | 10μg |
Source | Mouse myeloma cell line,NS0-derived human Angiopoietin-2 protein Tyr19-Phe496,with a C-terminal 6-his tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
Gene Alias | AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, ANGPT2, Tie2-ligand |
Common Name | Angiopoietin-2 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction