Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Angiopoietin-like 5 Protein

Catalog No. p-4719072
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human Angiopoietin-like Protein 5/ANGPTL5 (Catalog # 6675-AN/CF) is immobilized at 2 μg/mL (100 μL/well), Recombinant Human LILRB2/CD85d/ILT4 Fc Chimera (Catalog # 2078-T4 ) binds with an ED50 of 80.0-800 ng/mL.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Gene ID (Entrez) 253935
Molecular Weight (g/mol) 42.3 kDa
Name Angiopoietin-like Protein 5/ANGPTL5
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Angiopoietin-like Protein 5/ANGPTL5 protein Met1-Lys388 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.