Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Angiostatin Fc Chimera Protein
Click to view available options
Quantity:
50 μg
Description
Measured by the ability of the immobilized protein to support the adhesion of THP-1 human acute monocytic leukemia cells. Chavakis, T. et al. (2005) Blood 105:1036. The ED50 for this effect is 0.5-2.5 μg/mL.
Specifications
Specifications
| Accession Number | P00747 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 5340 |
| Molecular Weight (g/mol) | M.W. (Observed): 75-85 kDa, reducing conditions; M.W. (theoretical): 67.7 kDa (monomer) |
| Name | Angiostatin Fc Chimera |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived human Angiostatin protein Met-Asp Human IgG1 (Pro100-Lys330) IEGR Human Angiostatin (Kringle 1-4) (Val98-Ala459) (N-terminus) Accession # P00747 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction