Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Annexin V Protein
SDP

Catalog No. NBP130265 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130265 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130265 Supplier Novus Biologicals™ Supplier No. NBP130265
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

An un-tagged recombinant protein corresponding to the amino acids 1-320 of Human Annexin V The Recombinant Human Annexin V Protein is derived from E. coli. The Recombinant Human Annexin V Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P08758
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 1mM DTT, 10% glycerol
Gene ID (Entrez) 308
Molecular Weight (g/mol) 36kDa
Name Annexin V protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Annexin A5, 1-320aa, Human, E.coli (NP_001145.1).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.