Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human ApoE4 Protein
SDP

Catalog No. N19963410U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1000 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N19963410U 1000 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N19963410U Supplier Novus Biologicals™ Supplier No. NBP1996341000UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Western Blot, SDS-Page

Recombinant protein containing 300 amino acids for Human ApoE4

The Recombinant Human ApoE4 Protein is derived from E. coli. The Recombinant Human ApoE4 Protein has been validated for the following applications: Western Blot, SDS-Page.

Specifications

Concentration Lyoph
For Use With (Application) Western Blot, SDS-PAGE
Gene ID (Entrez) 348
Molecular Weight (g/mol) 34.4 kDa
Quantity 1000 μg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/ug
Gene Symbol APOE
Specific Reactivity ApoE4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute in dH2O to a concentration of 0.1 - 1.0 mg/mL. The solution can then be diluted into other aqueous buffers and stored at 4C for 1 week or -20°C for long-term storage.
Buffer Sterile filtered and lyophilized with no additives
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.