Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human Apolipoprotein A-I/ApoA1 Protein

Click to view available options
:
0.02mg; Unlabeled
Description
Recombinant bioactive protein containing 244 amino acids for Human Apolipoprotein A-I/ApoA1 The Recombinant Human Apolipoprotein A-I/ApoA1 Protein is derived from E. coli. The Recombinant Human Apolipoprotein A-I/ApoA1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Accession Number | NM_000039 |
Formulation | Sterile filtered and lyophilized with no additives. |
Gene ID (Entrez) | 335 |
Name | Apolipoprotein A-I/ApoA1 protein |
Purification Method | SDS-PAGE |
Storage Requirements | −20°C. Avoid freeze-thaw cycles. |
Gene Alias | APOA1, apoA-I, Apo-AI, Apolipoprotein A1, apolipoprotein A-I, MGC117399 |
Cross Reactivity | Human |
Purity or Quality Grade | >97%, by SDS-PAGE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction