Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human Apolipoprotein E/ApoE Protein

Click to view available options
:
0.1mg; Unlabeled
Description
Recombinant bioactive protein containing 299 amino acids for Human Apolipoprotein E/ApoE The Recombinant Human Apolipoprotein E/ApoE Protein is derived from E. coli. The Recombinant Human Apolipoprotein E/ApoE Protein has been validated for the following applications: Western Blot, SDS-Page.
Specifications
Specifications
Accession Number | NM_000041 |
Formulation | Lyophilized |
Gene ID (Entrez) | 348 |
Name | Apolipoprotein E/ApoE protein |
Storage Requirements | −20°C. Avoid freeze-thaw cycles. |
Cross Reactivity | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction