Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human ATPase Na+/K+ beta 2 His Protein
SDP

Catalog No. NB170696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170696 50 μg
NB170695 500 μg
NB170694 250 μg
NB170693 20 μg
NB170692 10 μg
NB170691 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. NB170696 Supplier Novus Biologicals™ Supplier No. NBP31511650UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specific activity is > 3,000 pmol/min/ug, and is defined as the amount of enzyme that hydrolyze 1.0 pmole of Adenosine 5-triphosphate to phosphate per minute per minute at pH 7.5 at 25C.

Specifications

Concentration 0.5 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 482
Molecular Weight (g/mol) M.W. Theoretical: 26.4 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <1 EU per 1ug of protein (determined by LAL method)
Gene Alias adhesion molecule on glia, AMOGsodium/potassium-dependent ATPase beta-2 subunit, ATPase, Na+/K+ transporting, beta 2 polypeptide, Na, K-ATPase beta-2 polypeptide, Sodium/potassium-dependent ATPase subunit beta-2, sodium/potassium-transporting ATPase beta-2 chain, sodium/potassium-transporting ATPase subunit beta-2
Gene Symbol ATP1B2
Research Category Endocrinology, Signal Transduction
Purity or Quality Grade >90%, by SDS-PAGE
Protein ATPase Na+/K+ beta 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.