Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human B7-1/CD80 Fc Chimera Protein
Click to view available options
Quantity:
1 mg
100 μg
Description
Measured by its ability to induce IL-2 secretion by Jurkat human acute T cell leukemia cells. Freeman, G.J. et al. (1993) Science 262:909. The ED50 for this effect is 0.025-0.15 μg/mL in the presence of PHA.
Specifications
Specifications
Accession Number | P33681 |
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
Gene ID (Entrez) | 941 |
Molecular Weight (g/mol) | M.W. (Observed): 66-80 kDa, under reducing conditions; M.W. (theoretical): 50.5 kDa |
Purification Method | >95%, by SDS-PAGE under reducing conditions and visualized by silver stain |
Quantity | 100 μg |
Source | Chinese Hamster Ovary cell line, CHO-derived human B7-1/CD80 protein Human B7-1/CD80 (Val35-Asn242) (N-terminus) Accession # P33681 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction