Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BDNF Protein

Catalog No. p-7161292
Click to view available options
Quantity:
1 mg
10 μg
250 μg
5 μg
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human BDNF Protein is derived from Sf 21 (baculovirus). The Recombinant Human BDNF Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P23560
For Use With (Application) Bioactivity
Gene ID (Entrez) 627
Molecular Weight (g/mol) 13-14 kDa, under reducing conditions.
Quantity 5 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived BDNF protein His129-Arg247 Accession &Num; P23560 100% sequence homology with Mouse,Rat,Canine,Equine and all other mammalian proteins examined.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Specific Reactivity BDNF
Conjugate Unconjugated
Species Human, Mouse, Rat, Canine, Equine
Recombinant Recombinant
Reconstitution Reconstitute 5 μg vials at 100 μg/mL in water. Reconstitute 10 μg or larger vials at 250 μg/mL in water.
Buffer Lyophilized from a 0.2 μm filtered solution in Sodium Citrate and NaCl with BSA as a carrier protein.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.