Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human beta-1,3-Glucuronyltransferase 1/B3GAT1 hIgG-His Protein
SDP

Catalog No. NB002933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.1 mg
0.5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002933 0.5 mg
NB002931 0.05 mg
NB002932 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB002933 Supplier Novus Biologicals™ Supplier No. NBP2774890.5MG

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a hIgG-His-Tag and corresponding to the amino acids 28-334aa of Human B3GAT1.

Source: Baculovirus

Amino Acid Sequence:

ADP>TLAPLLA VHKDEGSDPR RETPPGADPR EYCTSDRDIV EVVRTEYVYT RPPPWSDTLP TIHVVTPTYS RPVQKAELTR MANTLLHVPN LHWLVVEDAP RRTPLTARLL RDTGLNYTHL HVETPRNYKL RGDARDPRIP RGTMQRNLAL RWLRETFPRN SSQPGVVYFA DDDNTYSLEL FEEMRSTRRV SVWPVAFVGG LRYEAPRVNG AGKVVGWKTV FDPHRPFAID MAGFAVNLRL ILQRSQAYFK LRGVKGGYQE SSLLRELVTL NDLEPKAANC TKILVWHTRT EKPVLVNEGK KGFTDPSVEI LEPKSCDKTH TCPPCPAPEL LGGPSVFLFP PKPKDTLMIS RTPEVTCVVVDVSHEDPEVK FNWYVDGVEV HNAKTKPREE QYNSTYRVVS VLTVLHQDWL NGKEYKCKVSNKALPAPIEK TISKAKGQPR EPQVYTLPPS RDELTKNQVS LTCLVKGFYP SDIAVEWESNGQPENNYKTT PPVLDSDGSF FLYSKLTVDK SRWQQGNVFS CSVMHEALHNHYTQKSLSLS PGKHHHHHH>

Specifications

Concentration 0.25 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.5), 0.1M NaCl, 10% Glycerol
Gene ID (Entrez) 27087
Molecular Weight (g/mol) 62.3 kDa
Quantity 0.5 mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
Gene Symbol B3GAT1
Buffer PBS (pH 7.5), 0.1M NaCl, 10% Glycerol
Endotoxin Level < 1.0 EU per 1 μg of protein (determined by LAL method)
Purity or Quality Grade >90% SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.