Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human BMP-4 Protein
SDP

Catalog No. NB398399 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398399 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398399 Supplier Novus Biologicals™ Supplier No. NBP234931500UG
Add to Cart
Add to Cart

Recombinant Protein

Specifications

Accession Number P12644, P12644, P12644
Concentration LYOPH
For Use With (Application) Western Blot, ELISA, PAGE
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 50 mM Na2CO3, 5 mM DTT, 0.02% Tween 20, pH 11.0.
Gene ID (Entrez) 652
Molecular Weight (g/mol) 13.3 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of BMP-4 as determined by LAL method.
Gene Alias BMP-2B, BMP2B1, BMP2BMCOPS6, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, OFC11, ZYME
Gene Symbol BMP4
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Inhibitor Properties 1. Molecular standard (Western, ELISA) in studying secreted BMP-4; 2. Preparing antibodies for BMP-4 monomer; 3. Molecule standard in detecting secreted BMP-4 in reduced SDS-PAGE.
Research Category Biologically Active Proteins, Stem Cell Markers, Stem Cell Signaling Pathway
Purity or Quality Grade >95% by SDS-PAGE
Protein BMP-4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.