Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human BTN2A1/Butyrophilin 2A1 Protein

Catalog No. 9058BT050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Learn More About B7-related Butryophilins as Immune Modulators View Webinar

Measured by its binding ability in a functional ELISA. When Recombinant Human BTN2A1 is immobilized at 1 μg/mL (100 μL/well), the concentration of Recombinant Human DC-SIGN/CD209 Fc Chimera (Catalog # 161-DC ) that produces 50% of the optimal binding response is approximately 0.1-0.5 μg/mL.

Specifications

Accession Number Q7KYR7
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 11120
Molecular Weight (g/mol) M.W. (Observed): 39-44 kDa, reducing conditions; M.W. (theoretical): 25 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human BTN2A1 protein Gln29-Ala248, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias BK14H9.1, BT2.1, BTF1, BTF1BT2.1butyrophilin BTF1, butyrophilin subfamily 2 member A1, butyrophilin, subfamily 2, member A1, DJ3E1.1, FLJ36567
Common Name BTN2A1
Gene Symbol BTN2A1
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human BTN2A1 is immobilized at 1μg/mL (100μL/well), the concentration of Recombinant Human DC-SIGN/CD209 Fc Chimera that produces 50% of the optimal binding response is approximately 0.1-0.5μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Expression System Human embryonic kidney cell, HEK293-derived Gln29-Ala248, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein BTN2A1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.