Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human C1GalT1/C1GalT1C1 Protein

Catalog No. 8659GT020
Click to view available options
Quantity:
20 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to transfer galactose from UDP-galactose to 4-nitrophenyl-alpha-D-galactosaminide. The specific activity is >2,750 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q9NS00 (C1GalT1) & Q96EU7 (C1GalT1C1)
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 56913
Molecular Weight (g/mol) M.W. (Observed): 25-42 kDa, reducing conditions; M.W. (theoretical): 38 kDa (C1GalT1) & 35 kDa (C1GalT1C1)
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human C1GalT1 protein Asp45-Pro363, with N-terminal 6-His tag (C1GalT1); Ile28-Asp318, with an N-terminal HA tag (C1GalT1C1)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias B3Gal-T8, Beta-1,3-galactosyltransferase, C1GALT, C1GalT1, C1GalT2, C38H2-L1, Core 1 beta1,3-galactosyltransferase 1, Core 1 Beta3-Gal-T1, Core 1 O-Glycan T-Synthase, core 1 synthase, glycoprotein-N-acetylgalactosamine3-beta-galactosyltransferase, 1, Core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta1,3-galactosyltransferase 1, COSMC, EC 2.4.1.122, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1, MST143, TNPS, T-synthase
Common Name C1GALT1
Gene Symbol C1GALT1
Biological Activity Measured by its ability to transfer galactose from UDP-galactose to 4-nitrophenyl-alpha-D-galactosaminide. The specific activity is >2,750 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Asp45-Pro363, with N-terminal 6-His tag (C1GalT1); Ile28-Asp318, with an N-terminal HA tag (C1GalT1C1)
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein C1GALT1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.