Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Cardiotrophin-1/CT-1 Protein
SDP

Catalog No. NB002059 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002059 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002059 Supplier Novus Biologicals™ Supplier No. NBP19974550UG

Highly purified. Generating reliable and reproducible results.

Recombinant protein containing 200 amino acids for Human Cardiotrophin-1/CT-1 The Recombinant Human Cardiotrophin-1/CT-1 Protein is derived from E. coli. The Recombinant Human Cardiotrophin-1/CT-1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration Lyoph
For Use With (Application) SDS-PAGE
Formulation Sterile filtered and lyophilized from 1X PBS.
Gene ID (Entrez) 1489
Molecular Weight (g/mol) 22.5 kDa
Quantity 50 μg
Source E. coli
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias cardiophin 1, cardiotrophin 1, cardiotrophin-1, CT-1CT1
Gene Symbol CTF1
Reconstitution Reconstitute in 20 mM Tris, pH 8.0 to a concentration of <1.0 mg/mL. The solution can be diluted into other aqueous buffers.
Buffer Sterile filtered and lyophilized from 1X PBS.
Endotoxin Level <0.1 ng/μg
Purity or Quality Grade >98%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.