Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Carnosine Dipeptidase 1/CNDP1 His Protein
SDP

Catalog No. NBP25222705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25222705 0.05 mg
NBP25222725 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25222705 Supplier Novus Biologicals™ Supplier No. NBP2522270.05MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 27-507 of Human Carnosine Dipeptidase 1/CNDP1 The Recombinant Human Carnosine Dipeptidase 1/CNDP1 Protein is derived from Baculovirus. The Recombinant Human Carnosine Dipeptidase 1/CNDP1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 84735
Molecular Weight (g/mol) TMW: 54.9kDa
Quantity 0.05 mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase
Common Name Carnosine Dipeptidase 1/CNDP1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.