Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Casein Kinase 2 alpha Protein
Click to view available options
Quantity:
20 μg
Description
The Recombinant Human Casein Kinase 2 alpha Protein is derived from E. coli.
Specifications
Specifications
Accession Number | P68400-1 |
Formulation | Supplied as a 0.2 μm filtered solution in MES, NaCl. |
Gene ID (Entrez) | 1457 |
Molecular Weight (g/mol) | M.W. (Observed): 14-16 kDa, reducing conditions; M.W. (theoretical): 16 kDa |
Quantity | 20 μg |
Source | E. coli-derived human Casein Kinase 2 alpha protein Asp253-Gln391, with an N-terminal Met and C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
Regulatory Status | RUO |
Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction