Learn More
Description
Specifications
Specifications
| Concentration | 0.25 mg/ml |
| For Use With (Application) | Enzyme Activity, Functional Assay |
| Formulation | PBS (pH 7.4), 40% glycerol |
| Gene ID (Entrez) | 8722 |
| Molecular Weight (g/mol) | 52.5 kDa |
| Quantity | 0.05 mg |
| Storage Requirements | Store at -20°C short term. Aliquot and store at -70°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | < 1 EU per 1ug of protein (determined by LAL method) |
| Gene Alias | cathepsin F, CATSFEC 3.4.22.41, EC 3.4.22 |
| Gene Symbol | CTSF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
