Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Cathepsin L Protein
SDP

Catalog No. NB002054 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002054 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002054 Supplier Novus Biologicals™ Supplier No. NBP199623100UG

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant bioactive protein for Human Cathepsin L The Recombinant Human Cathepsin L Protein is derived from E. coli. The Recombinant Human Cathepsin L Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration 0.5 mg/mL
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 25 mM Sodium Acetate, 50 mM NaCl, pH 5.5 containing 50% glycerol
Gene ID (Entrez) 1514
Molecular Weight (g/mol) 24.2 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias cathepsin L, cathepsin L1, CATL, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP
Gene Symbol CTSL
Buffer 25 mM Sodium Acetate, 50 mM NaCl, pH 5.5 containing 50% glycerol
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.