Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CD20 Protein
SDP

Catalog No. p-7145873
Click to view available options
Quantity:
10 μg
50 μg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-terminalHis-tag and corresponding to the amino acids 210-297 of Human CD20. The Recombinant Human CD20 Protein is derived from Baculovirus. The Recombinant Human CD20 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 931
Molecular Weight (g/mol) 11.1 kDa
Quantity 10 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol MS4A1
Cross Reactivity CD20
Species Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.