Learn More
Description
Specifications
Specifications
| Accession Number | P10747, P10747, P10747, P10747, P10747 |
| Concentration | 1 mg/ml |
| For Use With (Application) | Binding Activity, Bioactivity, ELISA |
| Formulation | PBS (pH 7.4), 10% glycerol |
| Gene ID (Entrez) | 940 |
| Molecular Weight (g/mol) | M.W.-theoretical: 42.4 kDa |
| Quantity | 0.250 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <1.0 EU / 1 ug of protein (determined by LAL method) |
| Gene Alias | CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
