Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CD46 Protein
SDP

Catalog No. NB25304850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB25304850 50 μg
NB253048250 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB25304850 Supplier Novus Biologicals™ Supplier No. NBP25304850UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 35-313 of Human CD46 The Recombinant Human CD46 Protein is derived from Baculovirus. The Recombinant Human CD46 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 4179
Molecular Weight (g/mol) 32.5kDa
Name Human CD46 Protein
Purification Method >90%, by SDS-PAGE
Quantity 50 μg
Source Baculovirus
Immunogen ADLCEEPPTF EAMELIGKPK PYYEIGERVD YKCKKGYFYI PPLATHTICD RNHTWLPVSD DACYRETCPY IRDPLNGQAV PANGTYEFGY QMHFICNEGY YLIGEEILYC ELKGSVAIWS GKPPICEKVL CTPPPKIKNG KHTFSEVEVF EYLDAVTYSC DPAPGPDPFS LIGESTIYCG DNSVWSRAAP ECKVVKCRFP VVENGKQISG FGKKFYYKAT VMFECDKGFY LDGSDTIVCD SNSTWDPPVP KCLKGPRPTY KPPVSNYPGY PKPEEGILDS LDHHHHHH
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen
Gene Symbol CD46
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.