Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CD98 Protein
SDP

Catalog No. NB261178250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB261178250 250 μg
NBP26117850 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB261178250 Supplier Novus Biologicals™ Supplier No. NBP261178250UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 206-630 of Human CD98 The Recombinant Human CD98 Protein is derived from E. coli. The Recombinant Human CD98 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.5) containing 30% glycerol, 0.1M NaCl, 0.1mM PMSF
Gene ID (Entrez) 6520
Molecular Weight (g/mol) 47.9 kDa
Quantity 250 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1μg of protein (determined by LAL method)
Gene Symbol SLC3A2
Cross Reactivity CD98
Species Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.