Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human CHMP1B His Protein

Click to view available options
:
20μg; Unlabeled
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-199 of Human RSG1 The Recombinant Human RSG1 Protein is derived from E. coli. The Recombinant Human RSG1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Accession Number | Q9BZJ3 |
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. 20mM Tris-HCl buffer (pH8.0) containing 20% glycerol, 0.1M NaCl, 1mM DTT |
Gene ID (Entrez) | 23430 |
Molecular Weight (g/mol) | 24.7kDa |
Name | TPSD1 Protein |
Purification Method | SDS-PAGE |
Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
Cross Reactivity | Human |
Purity or Quality Grade | >85% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction