Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CHMP4A His Protein
SDP

Catalog No. NBP17887510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17887510 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17887510 Supplier Novus Biologicals™ Supplier No. NBP17887510UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-265 of Human CHMP4A The Recombinant Human CHMP4A Protein is derived from E. coli. The Recombinant Human CHMP4A Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q9BY43
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 2mM DTT, 50% glycerol, 200mM NaCl, 0.1mM PMSF, 1mM EDTA
Gene ID (Entrez) 29082
Molecular Weight (g/mol) 32kDa
Name CHMP4A Protein
Purification Method SDS-PAGE
Quantity 10 μg
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.