Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Chondromodulin-1/LECT1 His Protein
SDP

Catalog No. NBP25153905 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25153905 0.5 mg
NBP25153901 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25153905 Supplier Novus Biologicals™ Supplier No. NBP2515390.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 214-334 of Human Chondromodulin-1/LECT1 The Recombinant Human Chondromodulin-1/LECT1 Protein is derived from E. coli. The Recombinant Human Chondromodulin-1/LECT1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.4M UREA, 10% glycerol
Gene ID (Entrez) 11061
Molecular Weight (g/mol) TMW: 16.3kDa
Quantity 0.5 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias BRICD3, BRICHOS domain containing 3, CHM1chondromodulin I, CHMI, CHM-I, chondromodulin, chondromodulin-1, chondromodulin-I, leukocyte cell derived chemotaxin 1, Leukocyte cell-derived chemotaxin 1, multiple myeloma tumor suppressor 1, MYETS1
Common Name Chondromodulin-1/LECT1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.