Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CLECL1 GST (N-Term) Protein
SDP

Catalog No. NB398844 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398844 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398844 Supplier Novus Biologicals™ Supplier No. H00160365Q012UG

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 160365
Molecular Weight (g/mol) MolecularWeight-theroretical: 35.2 kDa
Quantity 2 μg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias C-type lectin-like 1, C-type lectin-like domain family 1, DCAL-1, DCAL1dendritic cell associated lectin 1, DC-associated lectin-1, Dendritic cell-associated lectin 1, dendritic cell-associated lectin-1, type II transmembrane protein DCAL1
Gene Symbol CLECL1
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein CLECL1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.