Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Complement Factor H-related 1 His-tag
Click to view available options
Quantity:
50 μg
Description
When Recombinant Human Complement Factor H-related 1/CFHR1 is immobilized at 1 μg/mL (100 μL/well), the concentration of Biotinylated Recombinant Mouse Complement Component C3d that produces 50% of the optimal binding response is 40 to 240 ng/mL.
Specifications
Specifications
Accession Number | Q03591 |
For Use With (Application) | Bioactivity |
Gene ID (Entrez) | 3078 |
Molecular Weight (g/mol) | 39 to 48 kDa, reducing conditions |
Quantity | 50 μg |
Source | Mouse myeloma cell line, NS0-derived human Complement Factor H-related 1/CFHR1 protein Glu19-Ala328, with a C-terminal 10-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
Endotoxin Concentration | < 0.10 EU per 1 μg of the protein by the LAL method |
Conjugate | Unconjugated |
Species | Human |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction