Learn More
Novus Biologicals™ Recombinant Human CXCL1/GRO alpha/KC/CINC-1 Protein
Shop All R&D Systems Products

Description
The Recombinant Human CXCL1/GRO alpha/KC/CINC-1 Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
| Concentration | Lyoph |
| For Use With (Application) | Bioactivity, SDS-PAGE |
| Gene ID (Entrez) | 2919 |
| Molecular Weight (g/mol) | 7.9 kDa |
| Purification Method | >95%, by SDS-PAGE |
| Quantity | 25 μg |
| Source | E. coli |
| Storage Requirements | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
| Gene Symbol | CXCL1 |
| Specific Reactivity | CXCL1 is a monomer, 7.9 kDa (73 aa) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.