Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human CXCL13/BLC/BCA-1 Protein
SDP

Catalog No. NB002056 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002056 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002056 Supplier Novus Biologicals™ Supplier No. NBP199658100UG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant bioactive protein containing 85 amino acids for Human CXCL13/BLC/BCA-1 The Recombinant Human CXCL13/BLC/BCA-1 Protein is derived from E. coli. The Recombinant Human CXCL13/BLC/BCA-1 Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Sterile filtered and lyophilized from 10 mM TFA (trifluoroacetic acid).
Gene ID (Entrez) 10563
Molecular Weight (g/mol) 10.1 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant)
Gene Symbol CXCL13
Reconstitution Centrifuge the vial prior to opening. Reconstitute in H2O to a concentration of 0.1 - 1 mg/mL. The solution can then be diluted into other aqueous buffers or stored at 4°C for 1 week or -20°C for long-term storage.
Buffer Sterile filtered and lyophilized from 10 mM TFA (trifluoroacetic acid).
Endotoxin Level <0.1 ng/μg
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.