Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Cytohesin 3 His Protein
SDP

Catalog No. NBP251672MG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP251672MG 0.02 mg
NBP25167101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP251672MG Supplier Novus Biologicals™ Supplier No. NBP2516710.02MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-399 of Human Cytohesin 3 The Recombinant Human Cytohesin 3 Protein is derived from E. coli. The Recombinant Human Cytohesin 3 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 9265
Molecular Weight (g/mol) TMW: 48.7kDa
Quantity 0.02 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias ARF nucleotide-binding site opener 3, ARNO3PH, SEC7 and coiled-coil domain-containing protein 3, cytohesin 3, General receptor of phosphoinositides 1, Grp1, GRP1cytohesin-3, pleckstrin homology, Sec7 and coiled-coil domains 3, Protein ARNO3, PSCD3pleckstrin homology, Sec7 and coiled/coil domains 3
Common Name Cytohesin 3
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.