Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human DLK2/EGFL9 Fc Protein
SDP

Catalog No. NB171703 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171703 50 μg
NB171702 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB171703 Supplier Novus Biologicals™ Supplier No. NBP31814550UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number Q6UY11
For Use With (Application) SDS-PAGE
Formulation Liquid. 0.2um-filtered solution in PBS
Gene ID (Entrez) 65989
Molecular Weight (g/mol) M.W. Theoretical: 70 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.1 EU/μg purified protein (LAL test; Lonza).
Gene Alias delta-like 2 homolog (Drosophila), DLK-2, EGFL9, EGF-like protein 9, EGF-like-domain, multiple 9, Epidermal growth factor-like protein 9, MGC111055, MGC2487, protein delta homolog 2
Gene Symbol DLK2
Research Category Cancer
Purity or Quality Grade >90% SDS-PAGE
Protein DLK2/EGFL9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.