Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human (DYKDDDDK) Ubiquitin Protein
Click to view available options
:
1mg; Carrier Free
Description
Recombinant Human (DYKDDDDK) Ubiquitin can be conjugated to substrate proteins via the subsequent actions of a Ubiquitin-activating (E1) enzyme, a Ubiquitin-conjugating (E2) enzyme, and a Ubiquitin ligase (E3). Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human (DYKDDDDK) Ubiquitin concentration of 0.5-1 mM.
Specifications
Specifications
Accession Number | P0CG47.1 |
Formulation | Lyophilized from a solution in deionized water. |
Gene ID (Entrez) | 7314 |
Molecular Weight (g/mol) | 9.8 kDa |
Name | FLAG Ubiquitin |
Quantity | 1mg |
Source | E. coli-derived human Ubiquitin protein Met1 - Gly76 with a N-terminal (DYKDDDDK) tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction