Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human EMAP-II/AIMP1 Protein
SDP

Catalog No. NB002028 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002028 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002028 Supplier Novus Biologicals™ Supplier No. NBP19938220UG

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant bioactive protein containing 166 amino acids for Human EMAP-II/AIMP1 The Recombinant Human EMAP-II/AIMP1 Protein is derived from E. coli. The Recombinant Human EMAP-II/AIMP1 Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Lyophilized from 20 mM sodium Phosphate buffer (pH 7.5), 130 mM NaCl.
Gene ID (Entrez) 9255
Molecular Weight (g/mol) 18.3 kDa
Quantity 20 μg
Source E. coli
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43
Gene Symbol AIMP1
Reconstitution Reconstitute in dH2O to a concentration of 0.1 - 1.0 mg/mL. The solution can then be diluted into other aqueous buffers and stored at 4°C for 1 week or -20°C for long-term storage.
Buffer Lyophilized from 20 mM sodium Phosphate buffer (pH 7.5), 130 mM NaCl.
Endotoxin Level <0.1 ng/μg
Purity or Quality Grade >98%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.