Learn More
Novus Biologicals Recombinant Human ERR beta/NR3B2 GST (N-Term) Protein
Shop All R&D Systems Products

Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, Protein Array |
| Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 2103 |
| Molecular Weight (g/mol) | 81.4 kDa |
| Quantity | 2 μg |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Gene Alias | estrogen-related receptor beta |
| Gene Symbol | ESRRB |
| Research Category | Cancer |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.