Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human FABP3/H-FABP Protein
SDP

Catalog No. NB398106 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398106 1 mg
NB398104 5 μg
NB398105 500 μg
NB398107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. NB398106 Supplier Novus Biologicals™ Supplier No. NBP3071071MG
Add to Cart
Add to Cart

Recombinant Protein

Specifications

For Use With (Application) SDS-PAGE
Formulation Lyophilized from a 0.2 mm filtered concentrated solution in 10 mM PB,150 mM NaCl,1 mM DTT, 0.1% Tween-20, pH 7.4.
Gene ID (Entrez) 2170
Molecular Weight (g/mol) MolecularWeight-theroretical: 14.7 kDa
Quantity 1 mg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1.0 EU/μg of FABP3/H-FABP as determined by LAL method.
Gene Alias FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP
Gene Symbol FABP3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Briefly centrifuge vial prior to opening to bring the contents to the bottom. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to 0.1-1.0 mg/mL Stock solutions should be apportioned into working aliquots and stored (at -20°C.)
Form Purified
Research Category Cell Cycle and Replication, Cholesterol Metabolism, Lipid and Metabolism
Purity or Quality Grade >95%, by SDS-PAGE
Protein FABP3/H-FABP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.