Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human FARP2 GST (N-Term) Protein
SDP

Catalog No. NB398893
Click to view available options
Quantity:
2 μg

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 9855
Molecular Weight (g/mol) 99.7 kDa
Quantity 2 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias FERM, RhoGEF and pleckstrin domain protein 2, FGD1-related Cdc42-GEF, FIRFERM domain including RhoGEF, FRG, KIAA0793PH domain-containing family C member 3, Pleckstrin homology domain-containing family C member 3, PLEKHC3FERM, RhoGEF and pleckstrin domain-containing protein 2
Gene Symbol FARP2
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein FARP2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.