Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FGF-22 Protein

Catalog No. p-4719651
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MES, Na 2 SO 4 EDTA, Sucrose and DTT with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in MES, Na 2 SO 4 EDTA, Sucrose and DTT.
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human FGF-22 Protein is derived from E. coli. The Recombinant Human FGF-22 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in MES, Na 2 SO 4 EDTA, Sucrose and DTT.
Gene ID (Entrez) 27006
Molecular Weight (g/mol) 18 kDa
Name FGF-22
Quantity 25 μg
Source E. coli-derived human FGF-22 protein Thr23-Ser170,with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.